The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142350.1 | NP_001135822.1 | 1 - 2 | 334 | 95 |
NM_024339.3 | NP_077315.2 | 1 - 2 | 334 | 95 | |
GenBank mRNA | AK075330.1 | - | 1 - 2 | 301 | 95 |
AK308960.1 | - | 1 - 2 | 322 | 95 | |
AK310113.1 | - | 1 - 2 | 334 | 95 | |
AK314086.1 | - | 1 - 2 | 312 | 95 | |
BC050674.1 | - | 1 - 2 | 273 | 95 | |
BG613317.1 | - | 1 - 2 | 335 | 95 |