The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032152.4 | NP_115528.4 | 2 - 3 | 1493 | 95 |
XM_005272502.2 | XP_005272559.1 | 2 - 3 | 1479 | 95 | |
XM_011528352.2 | XP_011526654.1 | 2 - 3 | 1499 | 95 | |
XM_011528353.2 | XP_011526655.1 | 2 - 3 | 1497 | 95 | |
GenBank mRNA | AJ272324.1 | - | 1 - 2 | 1637 | 95 |
AK308227.1 | - | 2 - 3 | 1476 | 95 | |
BC028012.1 | - | 3 - 4 | 1450 | 95 |