The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083113.1 | NP_001076582.1 | 3 - 4 | 869 | 105 |
NM_001195653.1 | NP_001182582.1 | 3 - 4 | 893 | 105 | |
NM_001195654.1 | NP_001182583.1 | 3 - 4 | 912 | 105 | |
NM_032527.4 | NP_115916.3 | 3 - 4 | 912 | 105 | |
NM_181485.2 | NP_852150.2 | 3 - 4 | 842 | 105 | |
GenBank mRNA | AB058750.3 | - | 3 - 4 | 846 | 105 |
AK027878.1 | - | 3 - 4 | 850 | 105 | |
AK074961.1 | - | 3 - 4 | 831 | 105 | |
AK314726.1 | - | 3 - 4 | 882 | 105 | |
BC019338.1 | - | 3 - 4 | 887 | 105 | |
BC032612.1 | - | 3 - 4 | 832 | 105 | |
KU178806.1 | - | 2 - 3 | 723 | 105 | |
KU178807.1 | - | 2 - 3 | 723 | 105 |