The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145862.1 | NP_001139334.1 | 4 - 5 | 576 | 56 |
NM_181873.3 | NP_870988.2 | 3 - 4 | 318 | 56 | |
XM_006711137.1 | XP_006711200.1 | 3 - 4 | 295 | 56 | |
XM_011509099.2 | XP_011507401.1 | 4 - 5 | 469 | 56 | |
GenBank mRNA | AK097000.1 | - | 3 - 4 | 295 | 56 |
AK296094.1 | - | 3 - 4 | 415 | 56 | |
AK298775.1 | - | 4 - 5 | 545 | 56 | |
AK304820.1 | - | 4 - 5 | 546 | 56 | |
BC032655.1 | - | 3 - 4 | 257 | 56 | |
BC064372.1 | - | 4 - 5 | 548 | 56 | |
BC094756.1 | - | 4 - 5 | 557 | 56 | |
BC100818.2 | - | 3 - 4 | 236 | 56 | |
BC100819.2 | - | 3 - 4 | 236 | 56 | |
BC100820.1 | - | 3 - 4 | 236 | 56 | |
BC100821.2 | - | 3 - 4 | 236 | 56 | |
DB474756.2 | - | 4 - 5 | 580 | 56 | |
U78556.1 | - | 4 - 5 | 511 | 56 | |
U78557.1 | - | 4 - 5 | 511 | 56 |