The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145862.1 | NP_001139334.1 | 6 - 7 | 800 | 86 |
NM_181873.3 | NP_870988.2 | 5 - 6 | 542 | 86 | |
XM_006711137.1 | XP_006711200.1 | 5 - 6 | 519 | 86 | |
XM_011509099.2 | XP_011507401.1 | 6 - 7 | 693 | 86 | |
GenBank mRNA | AK097000.1 | - | 5 - 6 | 519 | 86 |
AK296094.1 | - | 5 - 6 | 639 | 86 | |
AK298775.1 | - | 6 - 7 | 769 | 86 | |
AK304691.1 | - | 5 - 6 | 616 | 86 | |
AK304820.1 | - | 6 - 7 | 770 | 86 | |
BC032655.1 | - | 5 - 6 | 481 | 86 | |
BC064372.1 | - | 6 - 7 | 772 | 86 | |
BC094756.1 | - | 6 - 7 | 781 | 86 | |
BC100818.2 | - | 5 - 6 | 460 | 86 | |
BC100819.2 | - | 5 - 6 | 460 | 86 | |
BC100820.1 | - | 5 - 6 | 460 | 86 | |
BC100821.2 | - | 5 - 6 | 460 | 86 | |
U78556.1 | - | 6 - 7 | 735 | 86 | |
U78557.1 | - | 6 - 7 | 735 | 86 |