Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32580132-32580381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
UniGene
Hs.534322
Gene Chromosome Location
Chr.6: 32578769-32589836 on Build GRCh38
Transcripts
NM_001243965.1 NM_002124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5715314 c.764-7T>C p.? Chr.6: 32580277-32580277 on Build GRCh38
3830216 c.785C>G p.T262R Chr.6: 32580249-32580249 on Build GRCh38
5611975 c.780G>A p.Q260Q Chr.6: 32580254-32580254 on Build GRCh38
3684275 c.766C>A p.H256N Chr.6: 32580268-32580268 on Build GRCh38
4006423 c.777T>C p.L259L Chr.6: 32580257-32580257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758488110 Chr.6: 32580194-32580194 on Build GRCh38
rs770355132 Chr.6: 32580201-32580201 on Build GRCh38
rs747234648 Chr.6: 32580197-32580197 on Build GRCh38
rs17887154 Chr.6: 32580255-32580255 on Build GRCh38
rs760304327 Chr.6: 32580203-32580203 on Build GRCh38
rs777965283 Chr.6: 32580195-32580195 on Build GRCh38
rs28732249 Chr.6: 32580234-32580234 on Build GRCh38
rs779998278 Chr.6: 32580188-32580188 on Build GRCh38
rs771219818 Chr.6: 32580308-32580309 on Build GRCh38
rs36048298 Chr.6: 32580344-32580344 on Build GRCh38
rs777426966 Chr.6: 32580294-32580303 on Build GRCh38
rs149860663 Chr.6: 32580231-32580231 on Build GRCh38
rs9269741 Chr.6: 32580170-32580170 on Build GRCh38
rs766029790 Chr.6: 32580203-32580204 on Build GRCh38
rs4498414 Chr.6: 32580291-32580291 on Build GRCh38
rs746271300 Chr.6: 32580200-32580200 on Build GRCh38
rs781570813 Chr.6: 32580309-32580310 on Build GRCh38
rs752708985 Chr.6: 32580193-32580193 on Build GRCh38
rs41287243 Chr.6: 32580343-32580343 on Build GRCh38
rs761425415 Chr.6: 32580204-32580204 on Build GRCh38
rs35067512 Chr.6: 32580257-32580257 on Build GRCh38
rs551366042 Chr.6: 32580351-32580351 on Build GRCh38
rs201160908 Chr.6: 32580282-32580282 on Build GRCh38
rs28732257 Chr.6: 32580338-32580338 on Build GRCh38
rs9269743 Chr.6: 32580214-32580214 on Build GRCh38
rs4448132 Chr.6: 32580345-32580345 on Build GRCh38
rs28732256 Chr.6: 32580308-32580308 on Build GRCh38
rs772707468 Chr.6: 32580290-32580290 on Build GRCh38
rs746861116 Chr.6: 32580305-32580309 on Build GRCh38
rs149545032 Chr.6: 32580339-32580339 on Build GRCh38
rs200735393 Chr.6: 32580223-32580223 on Build GRCh38
rs772932482 Chr.6: 32580230-32580230 on Build GRCh38
rs28732255 Chr.6: 32580305-32580305 on Build GRCh38
rs775552150 Chr.6: 32580296-32580296 on Build GRCh38
rs41287241 Chr.6: 32580220-32580220 on Build GRCh38
rs199555742 Chr.6: 32580293-32580293 on Build GRCh38
rs141267330 Chr.6: 32580277-32580277 on Build GRCh38
rs28732251 Chr.6: 32580276-32580276 on Build GRCh38
rs35390341 Chr.6: 32580327-32580327 on Build GRCh38
rs759419167 Chr.6: 32580241-32580241 on Build GRCh38
rs192487435 Chr.6: 32580254-32580254 on Build GRCh38
rs35428228 Chr.6: 32580218-32580218 on Build GRCh38
rs41287239 Chr.6: 32580203-32580203 on Build GRCh38
rs28732254 Chr.6: 32580303-32580303 on Build GRCh38
rs71540436 Chr.6: 32580320-32580320 on Build GRCh38
rs773689016 Chr.6: 32580221-32580221 on Build GRCh38
rs73726893 Chr.6: 32580292-32580292 on Build GRCh38
rs142754652 Chr.6: 32580184-32580184 on Build GRCh38
rs41287240 Chr.6: 32580205-32580205 on Build GRCh38
rs9269742 Chr.6: 32580211-32580211 on Build GRCh38
rs28732248 Chr.6: 32580224-32580224 on Build GRCh38
rs572671781 Chr.6: 32580250-32580250 on Build GRCh38
rs199696659 Chr.6: 32580297-32580297 on Build GRCh38
rs780704359 Chr.6: 32580215-32580215 on Build GRCh38
rs28732252 Chr.6: 32580294-32580294 on Build GRCh38
rs767679220 Chr.6: 32580300-32580300 on Build GRCh38
rs376034726 Chr.6: 32580181-32580181 on Build GRCh38
rs776525968 Chr.6: 32580240-32580240 on Build GRCh38
rs9269740 Chr.6: 32580165-32580165 on Build GRCh38
rs755996563 Chr.6: 32580187-32580187 on Build GRCh38
rs757377940 Chr.6: 32580212-32580212 on Build GRCh38
rs753275248 Chr.6: 32580205-32580206 on Build GRCh38
rs28732253 Chr.6: 32580295-32580295 on Build GRCh38
rs545360810 Chr.6: 32580210-32580210 on Build GRCh38
rs34691126 Chr.6: 32580213-32580213 on Build GRCh38
rs796622039 Chr.6: 32580213-32580214 on Build GRCh38
rs796583582 Chr.6: 32580203-32580205 on Build GRCh38
rs756519999 Chr.6: 32580272-32580272 on Build GRCh38
rs762541895 Chr.6: 32580202-32580202 on Build GRCh38
rs760469607 Chr.6: 32580208-32580208 on Build GRCh38
rs9269745 Chr.6: 32580289-32580289 on Build GRCh38
rs9269746 Chr.6: 32580304-32580304 on Build GRCh38
rs3828830 Chr.6: 32580324-32580324 on Build GRCh38
rs41287242 Chr.6: 32580336-32580336 on Build GRCh38
rs9269747 Chr.6: 32580309-32580309 on Build GRCh38
rs749854667 Chr.6: 32580216-32580216 on Build GRCh38
rs771304919 Chr.6: 32580199-32580199 on Build GRCh38
rs752024157 Chr.6: 32580187-32580187 on Build GRCh38
rs778866014 Chr.6: 32580207-32580208 on Build GRCh38
rs531647126 Chr.6: 32580284-32580284 on Build GRCh38
rs781110703 Chr.6: 32580282-32580286 on Build GRCh38
rs374314091 Chr.6: 32580230-32580231 on Build GRCh38
rs199886031 Chr.6: 32580162-32580162 on Build GRCh38
rs28732250 Chr.6: 32580273-32580273 on Build GRCh38

View Full Product Details