The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014761.3 | NP_055576.2 | 8 - 9 | 964 | 151 |
GenBank mRNA | BC000430.2 | - | 8 - 9 | 914 | 151 |
BC004359.1 | - | 8 - 9 | 914 | 151 |