Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr.X: 49074433 - 49101121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029896.1 NP_001025067.1 1 - 2 157 157
GenBank mRNA AK023344.1 - 1 - 2 154 157
AK225122.1 - 1 - 2 136 157
AK301084.1 - 1 - 2 138 157
AK315717.1 - 1 - 2 141 157
BC000464.2 - 1 - 2 151 157
BC003037.1 - 1 - 2 151 157
BQ068083.1 - 1 - 2 157 157

View Full Product Details