The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138730.2 | NP_620058.1 | 5 - 6 | 400 | 154 |
GenBank mRNA | AF401520.1 | - | 5 - 6 | 291 | 154 |
BC009529.2 | - | 5 - 6 | 328 | 154 |