Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MTHFD2
- Entrez Gene ID
-
10797
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
- Gene Aliases
-
NMDMC
- Chromosome Location
-
Chr.2: 74198563 - 74215298 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027405.1 |
|
1 - 2 |
177 |
162 |
GenBank mRNA |
BC015062.1 |
- |
1 - 2 |
127 |
162 |
View Full Product Details