The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006014.4 | NP_006005.2 | 2 - 3 | 663 | 107 |
GenBank mRNA | BC015744.1 | - | 2 - 3 | 663 | 107 |
BC019012.2 | - | 2 - 3 | 285 | 107 | |
BC062330.1 | - | 2 - 3 | 376 | 107 | |
BQ649305.1 | - | 2 - 3 | 330 | 107 | |
X92896.1 | - | 2 - 3 | 317 | 107 |