The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080818.3 | NP_543008.3 | 4 - 4 | 1434 | 79 |
XM_005254043.3 | XP_005254100.1 | 4 - 4 | 1720 | 79 | |
XM_005254044.3 | XP_005254101.1 | 4 - 4 | 2741 | 79 | |
XM_005254045.2 | XP_005254102.1 | 4 - 4 | 1481 | 79 | |
XM_005254046.2 | XP_005254103.1 | 4 - 4 | 1571 | 79 | |
XM_011521077.2 | XP_011519379.1 | 4 - 4 | 1431 | 79 | |
GenBank mRNA | AJ305372.1 | - | 1 - 1 | 1207 | 79 |
AK122639.1 | - | 4 - 4 | 1434 | 79 |