The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002193.2 | 2 - 3 | 220 | 152 | |
GenBank mRNA | BC026077.2 | - | 2 - 3 | 222 | 152 |