Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EEF1A1
- Entrez Gene ID
-
1915
- Gene Name
- eukaryotic translation elongation factor 1 alpha 1
- Gene Aliases
-
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
- Chromosome Location
-
Chr.6: 73515750 - 73521032 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY043301.1 |
- |
1 - 1 |
270 |
147 |
View Full Product Details