The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144597.2 | NP_653198.2 | 4 - 4 | 886 | 149 |
GenBank mRNA | BC019820.1 | - | 4 - 4 | 851 | 149 |