Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDI1
Entrez Gene ID
414301
Gene Name
DNA damage inducible 1 homolog 1
Gene Aliases
-
Chromosome Location
Chr.11: 104036580 - 104039194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001711.2 NP_001001711.1 1 - 1 164 132
GenBank mRNA AF429971.1 - 1 - 1 160 132
AK093336.1 - 1 - 1 144 132
BC021172.2 - 1 - 1 105 132
BC021710.1 - 1 - 1 108 132
BC022017.2 - 1 - 1 131 132
BC022018.1 - 1 - 1 152 132
DB449163.1 - 1 - 1 165 132

View Full Product Details