Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123057721-123057982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6232875 c.1875T>C p.F625F Chr.11: 123057800-123057800 on Build GRCh38
3791279 c.1896C>T p.P632P Chr.11: 123057779-123057779 on Build GRCh38
5349630 c.1882G>C p.G628R Chr.11: 123057793-123057793 on Build GRCh38
6682660 c.1763A>G p.E588G Chr.11: 123057912-123057912 on Build GRCh38
924145 c.1911C>A p.S637S Chr.11: 123057764-123057764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946722582 Chr.11: 123057777-123057777 on Build GRCh38
rs367565153 Chr.11: 123057949-123057949 on Build GRCh38
rs118171965 Chr.11: 123057860-123057860 on Build GRCh38
rs563979190 Chr.11: 123057937-123057937 on Build GRCh38
rs371413585 Chr.11: 123057933-123057933 on Build GRCh38
rs758520548 Chr.11: 123057788-123057788 on Build GRCh38
rs756441145 Chr.11: 123057935-123057935 on Build GRCh38
rs112636139 Chr.11: 123057896-123057896 on Build GRCh38
rs749478600 Chr.11: 123057938-123057938 on Build GRCh38
rs370796977 Chr.11: 123057863-123057863 on Build GRCh38
rs760638348 Chr.11: 123057883-123057883 on Build GRCh38
rs780546314 Chr.11: 123057829-123057829 on Build GRCh38
rs987980360 Chr.11: 123057931-123057931 on Build GRCh38
rs144785833 Chr.11: 123057752-123057752 on Build GRCh38
rs768540563 Chr.11: 123057890-123057890 on Build GRCh38
rs755360656 Chr.11: 123057842-123057842 on Build GRCh38
rs781388678 Chr.11: 123057846-123057846 on Build GRCh38
rs776255814 Chr.11: 123057892-123057892 on Build GRCh38
rs183091882 Chr.11: 123057942-123057942 on Build GRCh38
rs753546825 Chr.11: 123057786-123057786 on Build GRCh38
rs757849635 Chr.11: 123057816-123057816 on Build GRCh38
rs762094637 Chr.11: 123057943-123057943 on Build GRCh38
rs764872570 Chr.11: 123057793-123057793 on Build GRCh38
rs41487746 Chr.11: 123057893-123057893 on Build GRCh38
rs781668860 Chr.11: 123057930-123057930 on Build GRCh38
rs201479266 Chr.11: 123057794-123057794 on Build GRCh38
rs997072349 Chr.11: 123057924-123057924 on Build GRCh38
rs377681653 Chr.11: 123057861-123057861 on Build GRCh38
rs4802 Chr.11: 123057914-123057914 on Build GRCh38
rs765426437 Chr.11: 123057816-123057827 on Build GRCh38
rs761588256 Chr.11: 123057902-123057902 on Build GRCh38
rs763753426 Chr.11: 123057779-123057779 on Build GRCh38
rs534622730 Chr.11: 123057924-123057924 on Build GRCh38
rs188408273 Chr.11: 123057851-123057851 on Build GRCh38
rs375617827 Chr.11: 123057925-123057925 on Build GRCh38
rs1056656311 Chr.11: 123057856-123057856 on Build GRCh38
rs3763897 Chr.11: 123057926-123057926 on Build GRCh38
rs775363477 Chr.11: 123057889-123057889 on Build GRCh38
rs755408420 Chr.11: 123057928-123057928 on Build GRCh38
rs145240324 Chr.11: 123057880-123057880 on Build GRCh38
rs780073872 Chr.11: 123057800-123057800 on Build GRCh38
rs776578052 Chr.11: 123057945-123057945 on Build GRCh38
rs963847272 Chr.11: 123057918-123057918 on Build GRCh38
rs776882071 Chr.11: 123057936-123057936 on Build GRCh38
rs533413759 Chr.11: 123057758-123057758 on Build GRCh38
rs752022565 Chr.11: 123057834-123057834 on Build GRCh38
rs750459503 Chr.11: 123057945-123057945 on Build GRCh38
rs779109914 Chr.11: 123057940-123057940 on Build GRCh38

View Full Product Details