The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098535.1 | NP_001092005.1 | 2 - 2 | 1224 | 85 |
NM_006604.2 | NP_006595.1 | 2 - 2 | 950 | 85 | |
GenBank mRNA | AJ010232.1 | - | 2 - 2 | 1224 | 85 |
AK097738.1 | - | 3 - 3 | 85 | ||
BC031689.1 | - | 2 - 2 | 950 | 85 |