Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
Chromosome Location
Chr.12: 52446651 - 52452126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005555.3 NP_005546.2 9 - 9 128
GenBank mRNA BC034535.1 - 9 - 9 1960 127

View Full Product Details