Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQCF2
Entrez Gene ID
389123
Gene Name
IQ motif containing F2
Gene Aliases
-
Chromosome Location
Chr.3: 51861629 - 51863424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203424.1 NP_982248.1 3 - 3 467 134
GenBank mRNA AK128883.1 - 3 - 3 508 134
BC040047.1 - 3 - 3 458 134

View Full Product Details