Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLRX5
Entrez Gene ID
51218
Gene Name
glutaredoxin 5
Gene Aliases
C14orf87, FLB4739, GRX5, PR01238, PRO1238, PRSA, SIDBA3, SPAHGC
Chromosome Location
Chr.14: 95534986 - 95544718 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016417.2 NP_057501.2 2 - 2 1007 135
GenBank mRNA AB223038.1 - 2 - 2 946 135
AF113691.1 - - 577 135
AI089763.1 - 1 - 1 135
BC047680.1 - 2 - 2 953 135
BC105275.1 - 1 - 1 484 135

View Full Product Details