The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031850.3 | NP_001027020.1 | 2 - 3 | 559 | 128 |
NM_002782.4 | NP_002773.1 | 2 - 3 | 559 | 128 | |
GenBank mRNA | BC020652.1 | - | 2 - 3 | 526 | 128 |
M31125.1 | - | 2 - 3 | 472 | 128 | |
M33666.1 | - | 2 - 3 | 479 | 128 | |
X17098.1 | - | 2 - 3 | 480 | 128 |