Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68208219-68208488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
UniGene
Hs.584921
Gene Chromosome Location
Chr.15: 68206992-68229742 on Build GRCh38
Transcripts
NM_017882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046117 c.721A>G p.M241V Chr.15: 68208355-68208355 on Build GRCh38
3503447 c.827G>A p.W276* Chr.15: 68208249-68208249 on Build GRCh38
6630566 c.795C>T p.S265S Chr.15: 68208281-68208281 on Build GRCh38
4649543 c.759C>G p.L253L Chr.15: 68208317-68208317 on Build GRCh38
3887364 c.798C>T p.F266F Chr.15: 68208278-68208278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs996513703 Chr.15: 68208450-68208451 on Build GRCh38
rs752910503 Chr.15: 68208271-68208271 on Build GRCh38
rs796052358 Chr.15: 68208370-68208370 on Build GRCh38
rs201306069 Chr.15: 68208455-68208455 on Build GRCh38
rs753994750 Chr.15: 68208355-68208355 on Build GRCh38
rs154774637 Chr.15: 68208364-68208364 on Build GRCh38
rs140653271 Chr.15: 68208335-68208335 on Build GRCh38
rs886186202 Chr.15: 68208390-68208390 on Build GRCh38
rs760271120 Chr.15: 68208308-68208308 on Build GRCh38
rs772090270 Chr.15: 68208362-68208362 on Build GRCh38
rs149262877 Chr.15: 68208353-68208353 on Build GRCh38
rs746753722 Chr.15: 68208397-68208397 on Build GRCh38
rs757481683 Chr.15: 68208356-68208356 on Build GRCh38
rs959199004 Chr.15: 68208376-68208376 on Build GRCh38
rs761161043 Chr.15: 68208413-68208413 on Build GRCh38
rs537994561 Chr.15: 68208326-68208326 on Build GRCh38
rs540227628 Chr.15: 68208344-68208344 on Build GRCh38
rs143781303 Chr.15: 68208309-68208309 on Build GRCh38
rs767164948 Chr.15: 68208348-68208348 on Build GRCh38
rs771007065 Chr.15: 68208282-68208282 on Build GRCh38
rs764363598 Chr.15: 68208424-68208424 on Build GRCh38
rs559620662 Chr.15: 68208406-68208406 on Build GRCh38
rs374681194 Chr.15: 68208321-68208321 on Build GRCh38
rs368443679 Chr.15: 68208404-68208404 on Build GRCh38
rs530677679 Chr.15: 68208465-68208465 on Build GRCh38
rs753344266 Chr.15: 68208245-68208245 on Build GRCh38
rs779052964 Chr.15: 68208284-68208284 on Build GRCh38
rs756310256 Chr.15: 68208276-68208276 on Build GRCh38
rs374553449 Chr.15: 68208444-68208444 on Build GRCh38
rs767889707 Chr.15: 68208336-68208336 on Build GRCh38
rs765433229 Chr.15: 68208441-68208441 on Build GRCh38
rs982013709 Chr.15: 68208363-68208363 on Build GRCh38
rs374744816 Chr.15: 68208398-68208398 on Build GRCh38
rs151295143 Chr.15: 68208307-68208307 on Build GRCh38
rs768422260 Chr.15: 68208280-68208282 on Build GRCh38
rs757531477 Chr.15: 68208432-68208432 on Build GRCh38
rs761419712 Chr.15: 68208400-68208400 on Build GRCh38
rs145746878 Chr.15: 68208322-68208322 on Build GRCh38
rs753014953 Chr.15: 68208411-68208411 on Build GRCh38
rs950362413 Chr.15: 68208328-68208328 on Build GRCh38
rs151186473 Chr.15: 68208254-68208254 on Build GRCh38
rs377192977 Chr.15: 68208343-68208343 on Build GRCh38
rs796052359 Chr.15: 68208267-68208267 on Build GRCh38
rs779750025 Chr.15: 68208366-68208366 on Build GRCh38
rs377098141 Chr.15: 68208431-68208431 on Build GRCh38
rs779105796 Chr.15: 68208358-68208358 on Build GRCh38
rs374613712 Chr.15: 68208277-68208277 on Build GRCh38
rs769339144 Chr.15: 68208401-68208401 on Build GRCh38
rs745850620 Chr.15: 68208359-68208359 on Build GRCh38
rs370531973 Chr.15: 68208458-68208458 on Build GRCh38
rs794729220 Chr.15: 68208239-68208247 on Build GRCh38
rs754129486 Chr.15: 68208430-68208430 on Build GRCh38
rs760345410 Chr.15: 68208243-68208243 on Build GRCh38
rs901756486 Chr.15: 68208275-68208275 on Build GRCh38
rs183399390 Chr.15: 68208266-68208266 on Build GRCh38
rs143728911 Chr.15: 68208244-68208244 on Build GRCh38
rs150363441 Chr.15: 68208301-68208301 on Build GRCh38
rs935136564 Chr.15: 68208452-68208452 on Build GRCh38
rs374271754 Chr.15: 68208407-68208407 on Build GRCh38
rs558758367 Chr.15: 68208287-68208287 on Build GRCh38
rs554594996 Chr.15: 68208327-68208327 on Build GRCh38
rs979072424 Chr.15: 68208443-68208443 on Build GRCh38
rs755397770 Chr.15: 68208262-68208262 on Build GRCh38
rs199634189 Chr.15: 68208453-68208453 on Build GRCh38
rs550705282 Chr.15: 68208466-68208466 on Build GRCh38
rs202012876 Chr.15: 68208255-68208255 on Build GRCh38
rs140319056 Chr.15: 68208278-68208278 on Build GRCh38

View Full Product Details