The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256355.1 | NP_001243284.1 | 4 - 4 | 2963 | 144 |
NM_001256356.1 | NP_001243285.1 | 3 - 3 | 2888 | 144 | |
NM_003347.3 | NP_003338.1 | 4 - 4 | 2984 | 144 | |
NR_028436.2 | 5 - 5 | 3093 | 144 | ||
NR_046082.1 | 4 - 4 | 3407 | 144 | ||
GenBank mRNA | AJ000519.1 | - | 4 - 4 | 2801 | 144 |
AK125396.1 | - | 4 - 4 | 3407 | 144 |