The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270942.1 | NP_001257871.1 | 5 - 5 | 1177 | 189 |
NM_001270943.1 | NP_001257872.1 | 4 - 4 | 1146 | 189 | |
NM_001270944.1 | NP_001257873.1 | 4 - 4 | 1110 | 189 | |
NM_003709.3 | NP_003700.1 | 4 - 4 | 1589 | 189 | |
NR_073108.1 | 5 - 5 | 983 | 189 | ||
XM_005246926.3 | XP_005246983.1 | 5 - 5 | 1378 | 189 | |
XM_006712815.3 | XP_006712878.1 | 5 - 5 | 1382 | 189 | |
XM_011512072.2 | XP_011510374.1 | 5 - 5 | 1556 | 189 | |
XM_011512073.2 | XP_011510375.1 | 6 - 6 | 1564 | 189 | |
XM_011512074.2 | XP_011510376.1 | 5 - 5 | 1159 | 189 | |
XM_017005161.1 | XP_016860650.1 | 5 - 5 | 1375 | 189 | |
XM_017005162.1 | XP_016860651.1 | 4 - 4 | 1128 | 189 | |
XM_017005164.1 | XP_016860653.1 | 4 - 4 | 1372 | 189 | |
GenBank mRNA | AB015132.1 | - | 4 - 4 | 1464 | 189 |
AK027371.1 | - | 4 - 4 | 1290 | 189 | |
AK297312.1 | - | 4 - 4 | 1099 | 191 | |
BC012919.1 | - | 5 - 5 | 1168 | 189 | |
BX537890.1 | - | 4 - 4 | 1148 | 191 |