Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL17
Entrez Gene ID
6139
Gene Name
ribosomal protein L17
Gene Aliases
L17, PD-1, RPL23
Chromosome Location
Chr.18: 49488481 - 49492565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000985.4 NP_000976.1 2 - 2 282 144
GenBank mRNA BC000502.2 - 1 - 1 185 144
BG497914.1 - 2 - 2 198 144

View Full Product Details