The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014062.2 | NP_054781.1 | 9 - 9 | 1622 | 153 |
NR_074074.1 | 8 - 8 | 1491 | 153 | ||
GenBank mRNA | AB026125.1 | - | 9 - 9 | 1580 | 153 |
AF190161.1 | - | 9 - 9 | 1549 | 153 | |
AK001028.1 | - | 9 - 9 | 1577 | 153 | |
AK126196.1 | - | 1 - 1 | 3107 | 153 | |
AY487344.1 | - | 9 - 9 | 1596 | 153 | |
BC064630.1 | - | 9 - 9 | 1565 | 153 | |
BM793847.1 | - | 1 - 1 | 306 | 153 | |
DB344693.1 | - | 1 - 1 | 153 |