Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10639366-10639609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_005263479.3 XM_017001539.1 XM_017001540.1 XM_017001541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4142076 c.4636G>C p.A1546P Chr.1: 10639586-10639586 on Build GRCh38
1199690 c.4810G>A p.A1604T Chr.1: 10639412-10639412 on Build GRCh38
7272679 c.4742C>T p.T1581M Chr.1: 10639480-10639480 on Build GRCh38
893272 c.4672G>A p.A1558T Chr.1: 10639550-10639550 on Build GRCh38
5989903 c.4835C>T p.P1612L Chr.1: 10639387-10639387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766290519 Chr.1: 10639416-10639416 on Build GRCh38
rs756224187 Chr.1: 10639484-10639484 on Build GRCh38
rs372231371 Chr.1: 10639503-10639503 on Build GRCh38
rs915304591 Chr.1: 10639547-10639547 on Build GRCh38
rs933919209 Chr.1: 10639410-10639410 on Build GRCh38
rs765922752 Chr.1: 10639551-10639551 on Build GRCh38
rs765233458 Chr.1: 10639573-10639573 on Build GRCh38
rs1004056195 Chr.1: 10639508-10639508 on Build GRCh38
rs749828128 Chr.1: 10639501-10639501 on Build GRCh38
rs760216629 Chr.1: 10639411-10639411 on Build GRCh38
rs749981678 Chr.1: 10639400-10639400 on Build GRCh38
rs752586999 Chr.1: 10639587-10639587 on Build GRCh38
rs539341512 Chr.1: 10639442-10639442 on Build GRCh38
rs757592574 Chr.1: 10639465-10639465 on Build GRCh38
rs753815288 Chr.1: 10639418-10639418 on Build GRCh38
rs376832173 Chr.1: 10639530-10639530 on Build GRCh38
rs754814432 Chr.1: 10639423-10639423 on Build GRCh38
rs774653511 Chr.1: 10639388-10639388 on Build GRCh38
rs988559517 Chr.1: 10639537-10639537 on Build GRCh38
rs779940295 Chr.1: 10639496-10639496 on Build GRCh38
rs765145459 Chr.1: 10639428-10639428 on Build GRCh38
rs758265341 Chr.1: 10639590-10639590 on Build GRCh38
rs761487730 Chr.1: 10639391-10639391 on Build GRCh38
rs775955020 Chr.1: 10639556-10639556 on Build GRCh38
rs772910102 Chr.1: 10639548-10639548 on Build GRCh38
rs781554397 Chr.1: 10639475-10639475 on Build GRCh38
rs767128575 Chr.1: 10639395-10639395 on Build GRCh38
rs982366563 Chr.1: 10639466-10639466 on Build GRCh38
rs995318030 Chr.1: 10639562-10639562 on Build GRCh38
rs746002511 Chr.1: 10639480-10639480 on Build GRCh38
rs771767807 Chr.1: 10639540-10639540 on Build GRCh38
rs889658581 Chr.1: 10639575-10639575 on Build GRCh38
rs760174926 Chr.1: 10639550-10639550 on Build GRCh38
rs558077733 Chr.1: 10639515-10639515 on Build GRCh38
rs759548779 Chr.1: 10639569-10639569 on Build GRCh38
rs1042882718 Chr.1: 10639560-10639560 on Build GRCh38
rs928221841 Chr.1: 10639467-10639467 on Build GRCh38
rs774453883 Chr.1: 10639567-10639567 on Build GRCh38
rs969602691 Chr.1: 10639441-10639441 on Build GRCh38
rs920916038 Chr.1: 10639398-10639398 on Build GRCh38

View Full Product Details