Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF5A
Entrez Gene ID
84844
Gene Name
PHD finger protein 5A
Gene Aliases
INI, Rds3, SAP14b, SF3B7, SF3b14b, bK223H9.2
Chromosome Location
Chr.22: 41459717 - 41468704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032758.3 NP_116147.1 4 - 4 918 145
GenBank mRNA BC007321.2 - 4 - 4 901 145
BC075808.1 - 4 - 4 887 145

View Full Product Details