The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193388.3 | NP_001180317.2 | 11 - 12 | 1383 | 120 |
NM_001193389.1 | NP_001180318.1 | 2 - 3 | 241 | 120 | |
NM_001276414.1 | NP_001263343.1 | 2 - 3 | 241 | 120 | |
XM_005255505.1 | XP_005255562.1 | 11 - 12 | 1412 | 120 | |
XM_011522610.1 | XP_011520912.1 | 12 - 13 | 1472 | 120 | |
XM_011522611.1 | XP_011520913.1 | 12 - 13 | 1472 | 120 | |
XM_011522612.1 | XP_011520914.1 | 11 - 12 | 1240 | 120 | |
XM_017023567.1 | XP_016879056.1 | 12 - 13 | 1472 | 120 | |
XM_017023568.1 | XP_016879057.1 | 2 - 3 | 238 | 120 | |
XM_017023569.1 | XP_016879058.1 | 2 - 3 | 353 | 120 | |
GenBank mRNA | AK027013.1 | - | 2 - 3 | 242 | 120 |
AM944365.1 | - | 11 - 12 | 1345 | 120 | |
BC000150.1 | - | 2 - 3 | 203 | 120 | |
DA558634.1 | - | 2 - 3 | 162 | 120 | |
DA739764.1 | - | 2 - 3 | 238 | 120 |