Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1QC
Entrez Gene ID
714
Gene Name
complement C1q C chain
Gene Aliases
C1Q-C, C1QG
Chromosome Location
Chr.1: 22643625 - 22648110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114101.1 NP_001107573.1 2 - 3 311 99
NM_172369.3 NP_758957.2 2 - 3 288 99
GenBank mRNA AF087892.1 - 2 - 3 286 99
AK057792.1 - 2 - 3 303 99
BC009016.1 - 2 - 3 253 99
CB995661.1 - 2 - 3 338 99
DA849505.1 - 2 - 3 280 99

View Full Product Details