The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007105.3 | NP_009036.2 | 3 - 4 | 1012 | 79 |
XM_017017833.1 | XP_016873322.1 | 3 - 4 | 2433 | 79 | |
GenBank mRNA | AF035407.1 | - | 1 - 2 | 379 | 79 |
AF037066.1 | - | 3 - 4 | 808 | 79 |