The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000109.3 | NP_000100.2 | 64 - 65 | 9681 | 78 |
NM_004006.2 | NP_003997.1 | 64 - 65 | 9605 | 78 | |
NM_004009.3 | NP_004000.1 | 64 - 65 | 9612 | 78 | |
NM_004010.3 | NP_004001.1 | 64 - 65 | 9695 | 78 | |
NM_004011.3 | NP_004002.2 | 36 - 37 | 5383 | 78 | |
NM_004012.3 | NP_004003.1 | 36 - 37 | 5526 | 78 | |
NM_004013.2 | NP_004004.1 | 21 - 22 | 3022 | 78 | |
NM_004014.2 | NP_004005.1 | 10 - 11 | 1235 | 78 | |
NM_004015.2 | NP_004006.1 | 3 - 4 | 235 | 78 | |
NM_004016.2 | NP_004007.1 | 3 - 4 | 235 | 78 | |
NM_004017.2 | NP_004008.1 | 3 - 4 | 235 | 78 | |
NM_004018.2 | NP_004009.1 | 3 - 4 | 235 | 78 | |
NM_004019.2 | NP_004010.1 | 3 - 4 | 235 | 78 | |
NM_004020.3 | NP_004011.2 | 21 - 22 | 3022 | 78 | |
NM_004021.2 | NP_004012.1 | 21 - 22 | 3022 | 78 | |
NM_004022.2 | NP_004013.1 | 21 - 22 | 3022 | 78 | |
NM_004023.2 | NP_004014.1 | 21 - 22 | 3022 | 78 | |
XM_006724468.2 | XP_006724531.1 | 64 - 65 | 9558 | 78 | |
XM_006724469.3 | XP_006724532.1 | 64 - 65 | 9532 | 78 | |
XM_006724470.3 | XP_006724533.1 | 64 - 65 | 9558 | 78 | |
XM_006724473.2 | XP_006724536.1 | 63 - 64 | 9420 | 78 | |
XM_006724474.3 | XP_006724537.1 | 64 - 65 | 9558 | 78 | |
XM_006724475.2 | XP_006724538.1 | 64 - 65 | 9558 | 78 | |
XM_011545467.1 | XP_011543769.1 | 63 - 64 | 9435 | 78 | |
XM_011545468.2 | XP_011543770.1 | 64 - 65 | 9558 | 78 | |
XM_017029328.1 | XP_016884817.1 | 64 - 65 | 9558 | 78 | |
XM_017029331.1 | XP_016884820.1 | 24 - 25 | 4134 | 78 | |
GenBank mRNA | AB208836.1 | - | 18 - 19 | 2469 | 78 |
AK299936.1 | - | 3 - 4 | 277 | 78 | |
BC028720.1 | - | 3 - 4 | 286 | 78 | |
BC070078.1 | - | 3 - 4 | 236 | 78 | |
BC094758.1 | - | 3 - 4 | 249 | 78 | |
BC127103.1 | - | 18 - 19 | 2546 | 78 | |
BC150141.1 | - | 18 - 19 | 2546 | 78 | |
DB233002.1 | - | 3 - 4 | 235 | 78 | |
KU178030.1 | - | 3 - 4 | 157 | 78 | |
KU178031.1 | - | 3 - 4 | 157 | 78 | |
KU178032.1 | - | 3 - 4 | 157 | 78 | |
KU178033.1 | - | 3 - 4 | 157 | 78 | |
M18533.1 | - | 64 - 65 | 9569 | 78 | |
M92650.1 | - | 3 - 4 | 209 | 78 | |
X14298.1 | - | 64 - 65 | 9459 | 78 |