Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
Chromosome Location
Chr.9: 27546546 - 27573866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256054.2 NP_001242983.1 3 - 4 704 143
NM_018325.4 NP_060795.1 3 - 4 609 143
NM_145005.6 NP_659442.2 3 - 4 626 143
GenBank mRNA AK057806.1 - 3 - 4 604 143
AK291425.1 - 3 - 4 599 143
AL832229.1 - 2 - 3 566 143
AL832467.1 - 3 - 4 565 143
BC068445.1 - 2 - 3 558 143
JN681271.1 - 3 - 4 682 143

View Full Product Details