The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073206.1 | 18 - 19 | 1856 | 115 | |
NR_073207.1 | 16 - 17 | 1737 | 115 | ||
XM_006723646.2 | XP_006723709.1 | 17 - 18 | 1737 | 115 | |
XM_006723647.2 | XP_006723710.1 | 17 - 18 | 1737 | 115 | |
XM_006723648.2 | XP_006723711.1 | 16 - 17 | 1680 | 115 | |
XM_006723654.3 | XP_006723717.1 | 8 - 9 | 752 | 115 | |
XM_011529374.1 | XP_011527676.1 | 16 - 17 | 1677 | 115 | |
XM_011529375.1 | XP_011527677.1 | 17 - 18 | 1626 | 115 | |
XM_011529376.1 | XP_011527678.1 | 16 - 17 | 1569 | 115 | |
XM_011529377.1 | XP_011527679.1 | 18 - 19 | 1946 | 115 | |
XM_011529378.1 | XP_011527680.1 | 15 - 16 | 1455 | 115 | |
XM_011529379.1 | XP_011527681.1 | 15 - 16 | 1488 | 115 | |
XM_017028084.1 | XP_016883573.1 | 6 - 7 | 674 | 115 | |
GenBank mRNA | AL137678.1 | - | 16 - 17 | 1621 | 115 |