The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308056.1 | NP_001294985.1 | 5 - 6 | 608 | 95 |
NM_001308059.1 | NP_001294988.1 | 6 - 7 | 643 | 95 | |
NM_001308060.1 | NP_001294989.1 | 3 - 4 | 373 | 95 | |
NM_001308061.1 | NP_001294990.1 | 4 - 5 | 500 | 95 | |
NM_139164.2 | NP_631903.1 | 5 - 6 | 614 | 95 | |
XM_017009041.1 | XP_016864530.1 | 4 - 5 | 425 | 95 | |
XM_017009042.1 | XP_016864531.1 | 4 - 5 | 419 | 95 | |
XM_017009043.1 | XP_016864532.1 | 3 - 4 | 337 | 95 | |
XM_017009044.1 | XP_016864533.1 | 4 - 5 | 407 | 95 | |
GenBank mRNA | AF480299.1 | - | 4 - 5 | 392 | 95 |
AK054566.1 | - | 5 - 6 | 476 | 95 | |
AK290312.1 | - | 5 - 6 | 527 | 95 | |
AK315863.1 | - | 3 - 4 | 311 | 95 | |
AK315869.1 | - | 4 - 5 | 425 | 95 | |
AL710253.1 | - | 5 - 6 | 573 | 95 | |
AL832599.2 | - | 1 - 2 | 70 | 95 |