Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGIP
Entrez Gene ID
492311
Gene Name
IgA inducing protein
Gene Aliases
C5orf53
Chromosome Location
Chr.5: 140125936 - 140128806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007189.1 NP_001007190.1 1 - 1 2571 137
GenBank mRNA AB073888.1 - 1 - 1 887 137
BC017422.1 - 1 - 1 163 137
BC041380.1 - 1 - 1 2575 137

View Full Product Details