Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INIP
Entrez Gene ID
58493
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C9orf80, HSPC043, MISE, SOSSC, SSBIP1, hSSBIP1
Chromosome Location
Chr.9: 112686506 - 112718107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021218.1 NP_067041.1 5 - 5 1124 113
GenBank mRNA AK000637.1 - 5 - 5 1127 113
BC014881.1 - 5 - 5 1124 113
BC065210.1 - 5 - 5 1122 113

View Full Product Details