Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MKRN1
Entrez Gene ID
23608
Gene Name
makorin ring finger protein 1
Gene Aliases
RNF61
Chromosome Location
Chr.7: 140453040 - 140479569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145125.1 NP_001138597.1 5 - 5 1581 119
NM_001291663.1 NP_001278592.1 7 - 7 1743 119
GenBank mRNA AF117233.1 - 5 - 5 1508 119
AM236048.1 - 5 - 5 1362 119
CA438497.1 - 1 - 1 119

View Full Product Details