The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020997.3 | NP_066277.1 | 4 - 4 | 1372 | 136 |
GenBank mRNA | AF081512.1 | - | 4 - 4 | 1362 | 136 |
AK129605.1 | - | 4 - 4 | 1075 | 136 | |
AY358873.1 | - | 4 - 4 | 1359 | 136 | |
BC027883.1 | - | 4 - 4 | 1359 | 136 |