Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92487420-92487645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
UniGene
Hs.164682
Gene Chromosome Location
Chr.7: 92487023-92528531 on Build GRCh38
Transcripts
NM_000466.2 NM_001282677.1 NM_001282678.1 XM_017012319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1452833 c.3824G>A p.R1275Q Chr.7: 92487485-92487485 on Build GRCh38
1093054 c.3772G>T p.E1258* Chr.7: 92487537-92487537 on Build GRCh38
1093053 c.3823C>T p.R1275* Chr.7: 92487486-92487486 on Build GRCh38
6432291 c.3788C>A p.P1263Q Chr.7: 92487521-92487521 on Build GRCh38
5501042 c.3768-4T>G p.? Chr.7: 92487545-92487545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765407359 Chr.7: 92487587-92487587 on Build GRCh38
rs1046054767 Chr.7: 92487593-92487593 on Build GRCh38
rs761960160 Chr.7: 92487580-92487580 on Build GRCh38
rs778216029 Chr.7: 92487554-92487555 on Build GRCh38
rs757816709 Chr.7: 92487587-92487591 on Build GRCh38
rs758748151 Chr.7: 92487500-92487501 on Build GRCh38
rs747271693 Chr.7: 92487569-92487569 on Build GRCh38
rs915545336 Chr.7: 92487585-92487585 on Build GRCh38
rs183618298 Chr.7: 92487605-92487605 on Build GRCh38
rs750667686 Chr.7: 92487592-92487592 on Build GRCh38
rs957039229 Chr.7: 92487458-92487458 on Build GRCh38
rs972848411 Chr.7: 92487606-92487606 on Build GRCh38
rs991200065 Chr.7: 92487561-92487561 on Build GRCh38
rs373346069 Chr.7: 92487473-92487473 on Build GRCh38
rs750722718 Chr.7: 92487477-92487477 on Build GRCh38
rs749050363 Chr.7: 92487537-92487537 on Build GRCh38
rs767554606 Chr.7: 92487562-92487562 on Build GRCh38
rs771765685 Chr.7: 92487559-92487560 on Build GRCh38
rs774248025 Chr.7: 92487557-92487557 on Build GRCh38
rs759357103 Chr.7: 92487560-92487560 on Build GRCh38
rs267608182 Chr.7: 92487459-92487459 on Build GRCh38
rs758795939 Chr.7: 92487478-92487478 on Build GRCh38
rs780491224 Chr.7: 92487483-92487483 on Build GRCh38
rs773001151 Chr.7: 92487527-92487527 on Build GRCh38
rs375758017 Chr.7: 92487508-92487508 on Build GRCh38
rs770868309 Chr.7: 92487542-92487542 on Build GRCh38
rs757410958 Chr.7: 92487455-92487455 on Build GRCh38
rs150793635 Chr.7: 92487485-92487485 on Build GRCh38
rs776520125 Chr.7: 92487573-92487573 on Build GRCh38
rs373360561 Chr.7: 92487568-92487568 on Build GRCh38
rs765142376 Chr.7: 92487470-92487470 on Build GRCh38
rs755549316 Chr.7: 92487486-92487486 on Build GRCh38
rs959756490 Chr.7: 92487516-92487516 on Build GRCh38
rs139054881 Chr.7: 92487499-92487499 on Build GRCh38

View Full Product Details