The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126.3 | NP_001117.2 | 9 - 10 | 1255 | 81 |
XM_011544108.2 | XP_011542410.1 | 9 - 10 | 1927 | 81 | |
GenBank mRNA | AK025514.1 | - | 9 - 10 | 1002 | 81 |
AK303886.1 | - | 10 - 11 | 967 | 81 | |
BC012356.1 | - | 9 - 10 | 1040 | 81 |