The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166139.2 | NP_001159611.1 | 3 - 4 | 421 | 105 |
NM_153686.8 | NP_710153.2 | 3 - 4 | 421 | 105 | |
XM_011513821.2 | XP_011512123.1 | 3 - 4 | 299 | 105 | |
XM_011513822.2 | XP_011512124.1 | 3 - 4 | 299 | 105 | |
XM_011513825.2 | XP_011512127.1 | 3 - 4 | 299 | 105 | |
XM_011513826.2 | XP_011512128.1 | 3 - 4 | 299 | 105 | |
XM_017007961.1 | XP_016863450.1 | 3 - 4 | 1833 | 105 | |
XM_017007964.1 | XP_016863453.1 | 3 - 4 | 250 | 105 | |
XM_017007965.1 | XP_016863454.1 | 3 - 4 | 1732 | 105 | |
GenBank mRNA | AK096181.1 | - | 4 - 5 | 282 | 105 |
BC037322.1 | - | 3 - 4 | 304 | 105 | |
BU902997.1 | - | 4 - 5 | 430 | 105 | |
DA408372.1 | - | 3 - 4 | 406 | 105 |