Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTMS
Entrez Gene ID
5763
Gene Name
parathymosin
Gene Aliases
ParaT
Chromosome Location
Chr.12: 6766139 - 6770952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002824.4 NP_002815.3 5 - 5 929 134
XM_011520987.1 XP_011519289.1 5 - 5 1242 134
GenBank mRNA BC017025.1 - 5 - 5 648 134
BE791815.1 - 2 - 2 379 134
BM451233.1 - 1 - 1 134
M24398.1 - 5 - 5 133

View Full Product Details