Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 96146485-96146758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A13
Entrez Gene ID
10165
Gene Name
solute carrier family 25 member 13
Gene Aliases
ARALAR2, CITRIN, CTLN2
UniGene
Hs.489190
Gene Chromosome Location
Chr.7: 96120220-96322147 on Build GRCh38
Transcripts
NM_001160210.1 NM_014251.2 NR_027662.1 XM_006715831.3 XM_011515727.2 XM_017011663.1 XM_017011664.1 XM_017011665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2865694 c.1363C>T p.R455C Chr.7: 96146645-96146645 on Build GRCh38
2865692 c.1372G>A p.V458M Chr.7: 96146636-96146636 on Build GRCh38
1698952 c.1342C>T p.P448S Chr.7: 96146666-96146666 on Build GRCh38
3730178 c.1364G>A p.R455H Chr.7: 96146644-96146644 on Build GRCh38
5014709 c.1406G>A p.S469N Chr.7: 96146602-96146602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576739220 Chr.7: 96146637-96146637 on Build GRCh38
rs771930914 Chr.7: 96146518-96146518 on Build GRCh38
rs143877538 Chr.7: 96146654-96146654 on Build GRCh38
rs115266882 Chr.7: 96146634-96146634 on Build GRCh38
rs745707033 Chr.7: 96146515-96146515 on Build GRCh38
rs367988218 Chr.7: 96146557-96146557 on Build GRCh38
rs897593798 Chr.7: 96146707-96146707 on Build GRCh38
rs780063972 Chr.7: 96146595-96146595 on Build GRCh38
rs780138239 Chr.7: 96146537-96146538 on Build GRCh38
rs761211929 Chr.7: 96146583-96146583 on Build GRCh38
rs778238353 Chr.7: 96146537-96146539 on Build GRCh38
rs771211154 Chr.7: 96146556-96146556 on Build GRCh38
rs749274731 Chr.7: 96146554-96146554 on Build GRCh38
rs372216502 Chr.7: 96146615-96146615 on Build GRCh38
rs879014162 Chr.7: 96146564-96146564 on Build GRCh38
rs1037190907 Chr.7: 96146664-96146664 on Build GRCh38
rs778898489 Chr.7: 96146734-96146734 on Build GRCh38
rs761402505 Chr.7: 96146645-96146645 on Build GRCh38
rs746775592 Chr.7: 96146600-96146600 on Build GRCh38
rs867546321 Chr.7: 96146736-96146736 on Build GRCh38
rs764951987 Chr.7: 96146592-96146592 on Build GRCh38
rs373629670 Chr.7: 96146688-96146688 on Build GRCh38
rs781202495 Chr.7: 96146686-96146686 on Build GRCh38
rs747368721 Chr.7: 96146602-96146602 on Build GRCh38
rs764488086 Chr.7: 96146584-96146584 on Build GRCh38
rs756072610 Chr.7: 96146719-96146719 on Build GRCh38
rs749506606 Chr.7: 96146537-96146537 on Build GRCh38
rs200237622 Chr.7: 96146672-96146672 on Build GRCh38
rs774369510 Chr.7: 96146563-96146563 on Build GRCh38
rs754985592 Chr.7: 96146608-96146608 on Build GRCh38
rs754531589 Chr.7: 96146536-96146537 on Build GRCh38
rs267601652 Chr.7: 96146655-96146655 on Build GRCh38
rs146111714 Chr.7: 96146574-96146574 on Build GRCh38
rs780158338 Chr.7: 96146520-96146520 on Build GRCh38
rs367766651 Chr.7: 96146721-96146721 on Build GRCh38
rs771217878 Chr.7: 96146708-96146708 on Build GRCh38
rs766077420 Chr.7: 96146668-96146668 on Build GRCh38
rs754659147 Chr.7: 96146685-96146685 on Build GRCh38
rs936911753 Chr.7: 96146667-96146667 on Build GRCh38
rs554809009 Chr.7: 96146588-96146588 on Build GRCh38
rs747004342 Chr.7: 96146531-96146531 on Build GRCh38
rs770856997 Chr.7: 96146616-96146616 on Build GRCh38
rs886062526 Chr.7: 96146695-96146695 on Build GRCh38
rs768713125 Chr.7: 96146537-96146537 on Build GRCh38
rs540149539 Chr.7: 96146609-96146609 on Build GRCh38
rs746153244 Chr.7: 96146620-96146620 on Build GRCh38
rs764693182 Chr.7: 96146644-96146644 on Build GRCh38
rs771592795 Chr.7: 96146546-96146547 on Build GRCh38
rs758167270 Chr.7: 96146660-96146660 on Build GRCh38
rs759905821 Chr.7: 96146570-96146570 on Build GRCh38
rs773624358 Chr.7: 96146589-96146592 on Build GRCh38
rs764716490 Chr.7: 96146649-96146649 on Build GRCh38
rs749990010 Chr.7: 96146593-96146593 on Build GRCh38
rs201598915 Chr.7: 96146589-96146589 on Build GRCh38
rs772435881 Chr.7: 96146738-96146738 on Build GRCh38
rs772250175 Chr.7: 96146628-96146628 on Build GRCh38
rs753411740 Chr.7: 96146585-96146585 on Build GRCh38
rs774476574 Chr.7: 96146617-96146617 on Build GRCh38
rs773388625 Chr.7: 96146538-96146538 on Build GRCh38
rs993631308 Chr.7: 96146709-96146709 on Build GRCh38

View Full Product Details