Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166199318-166199573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2 XM_011511616.2 XM_011511617.2 XM_011511618.2 XM_017004668.1 XM_017004669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7137067 c.5164C>A p.H1722N Chr.2: 166199478-166199478 on Build GRCh38
4571656 c.5121T>A p.P1707P Chr.2: 166199521-166199521 on Build GRCh38
6041339 c.5243C>A p.S1748Y Chr.2: 166199399-166199399 on Build GRCh38
4498181 c.5141C>T p.S1714L Chr.2: 166199465-166199465 on Build GRCh38
4582763 c.5167C>T p.P1723S Chr.2: 166199475-166199475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746833579 Chr.2: 166199512-166199512 on Build GRCh38
rs200817449 Chr.2: 166199419-166199419 on Build GRCh38
rs200637125 Chr.2: 166199546-166199546 on Build GRCh38
rs202208945 Chr.2: 166199452-166199452 on Build GRCh38
rs372887865 Chr.2: 166199478-166199478 on Build GRCh38
rs776586182 Chr.2: 166199432-166199432 on Build GRCh38
rs757406555 Chr.2: 166199363-166199364 on Build GRCh38
rs761637677 Chr.2: 166199437-166199437 on Build GRCh38
rs770424172 Chr.2: 166199401-166199401 on Build GRCh38
rs367959290 Chr.2: 166199361-166199361 on Build GRCh38
rs370267820 Chr.2: 166199530-166199530 on Build GRCh38
rs794727440 Chr.2: 166199404-166199404 on Build GRCh38
rs938958211 Chr.2: 166199498-166199498 on Build GRCh38
rs1013397100 Chr.2: 166199356-166199356 on Build GRCh38
rs200636760 Chr.2: 166199394-166199394 on Build GRCh38
rs200266625 Chr.2: 166199468-166199468 on Build GRCh38
rs376892319 Chr.2: 166199523-166199523 on Build GRCh38
rs199567943 Chr.2: 166199534-166199534 on Build GRCh38
rs200733722 Chr.2: 166199499-166199499 on Build GRCh38
rs267598971 Chr.2: 166199535-166199535 on Build GRCh38
rs768548713 Chr.2: 166199517-166199517 on Build GRCh38
rs778164639 Chr.2: 166199480-166199480 on Build GRCh38
rs201554947 Chr.2: 166199520-166199520 on Build GRCh38
rs775277644 Chr.2: 166199416-166199416 on Build GRCh38
rs1046237977 Chr.2: 166199358-166199358 on Build GRCh38
rs763972874 Chr.2: 166199427-166199427 on Build GRCh38
rs771718677 Chr.2: 166199500-166199500 on Build GRCh38
rs766522690 Chr.2: 166199441-166199441 on Build GRCh38
rs879253908 Chr.2: 166199388-166199388 on Build GRCh38
rs771700555 Chr.2: 166199413-166199413 on Build GRCh38
rs201980654 Chr.2: 166199386-166199386 on Build GRCh38
rs751682235 Chr.2: 166199442-166199442 on Build GRCh38
rs372210358 Chr.2: 166199392-166199392 on Build GRCh38
rs199806034 Chr.2: 166199487-166199487 on Build GRCh38
rs749848631 Chr.2: 166199482-166199482 on Build GRCh38
rs1027061303 Chr.2: 166199482-166199482 on Build GRCh38
rs201537698 Chr.2: 166199444-166199444 on Build GRCh38
rs201264153 Chr.2: 166199475-166199475 on Build GRCh38
rs755222805 Chr.2: 166199354-166199354 on Build GRCh38
rs759253467 Chr.2: 166199410-166199410 on Build GRCh38
rs761880427 Chr.2: 166199543-166199543 on Build GRCh38

View Full Product Details