Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 73168943-73169204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA1
Entrez Gene ID
301
Gene Name
annexin A1
Gene Aliases
ANX1, LPC1
UniGene
Hs.494173
Gene Chromosome Location
Chr.9: 73151731-73170393 on Build GRCh38
Transcripts
NM_000700.2 XM_011518609.1 XM_017014657.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5359307 c.900G>T p.M300I Chr.9: 73169070-73169070 on Build GRCh38
3699812 c.901G>A p.V301I Chr.9: 73169071-73169071 on Build GRCh38
7093047 c.937G>A p.A313T Chr.9: 73169107-73169107 on Build GRCh38
1625081 c.863G>T p.G288V Chr.9: 73169033-73169033 on Build GRCh38
3658035 c.893G>A p.R298K Chr.9: 73169063-73169063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756551545 Chr.9: 73169044-73169044 on Build GRCh38
rs761621511 Chr.9: 73169081-73169081 on Build GRCh38
rs781712414 Chr.9: 73169033-73169033 on Build GRCh38
rs550278720 Chr.9: 73168995-73168995 on Build GRCh38
rs750505719 Chr.9: 73169172-73169172 on Build GRCh38
rs746634649 Chr.9: 73169160-73169160 on Build GRCh38
rs762750858 Chr.9: 73169087-73169087 on Build GRCh38
rs780876153 Chr.9: 73169164-73169164 on Build GRCh38
rs1035662921 Chr.9: 73169060-73169060 on Build GRCh38
rs779798345 Chr.9: 73169056-73169056 on Build GRCh38
rs1025883774 Chr.9: 73169153-73169153 on Build GRCh38
rs749629349 Chr.9: 73169050-73169050 on Build GRCh38
rs753289028 Chr.9: 73169115-73169115 on Build GRCh38
rs769632738 Chr.9: 73169169-73169169 on Build GRCh38
rs867507521 Chr.9: 73169047-73169047 on Build GRCh38
rs756611110 Chr.9: 73169116-73169116 on Build GRCh38
rs769727131 Chr.9: 73169082-73169082 on Build GRCh38
rs764235820 Chr.9: 73169024-73169025 on Build GRCh38
rs776755767 Chr.9: 73169078-73169078 on Build GRCh38
rs570755224 Chr.9: 73169045-73169045 on Build GRCh38
rs751774481 Chr.9: 73169094-73169094 on Build GRCh38
rs375109829 Chr.9: 73169061-73169061 on Build GRCh38
rs139228721 Chr.9: 73169129-73169129 on Build GRCh38
rs766591114 Chr.9: 73169093-73169093 on Build GRCh38
rs779921203 Chr.9: 73169130-73169130 on Build GRCh38
rs867226678 Chr.9: 73169046-73169046 on Build GRCh38
rs370997349 Chr.9: 73169042-73169042 on Build GRCh38
rs751716730 Chr.9: 73169159-73169159 on Build GRCh38
rs771649725 Chr.9: 73169052-73169052 on Build GRCh38
rs767563984 Chr.9: 73169102-73169102 on Build GRCh38
rs757786869 Chr.9: 73169160-73169161 on Build GRCh38
rs747640646 Chr.9: 73169167-73169167 on Build GRCh38
rs759672522 Chr.9: 73169101-73169101 on Build GRCh38
rs773134758 Chr.9: 73169173-73169173 on Build GRCh38
rs768319087 Chr.9: 73169162-73169162 on Build GRCh38
rs778298033 Chr.9: 73169124-73169124 on Build GRCh38
rs953415008 Chr.9: 73169099-73169099 on Build GRCh38
rs746524290 Chr.9: 73169059-73169059 on Build GRCh38
rs751648003 Chr.9: 73169020-73169020 on Build GRCh38
rs539626064 Chr.9: 73169086-73169086 on Build GRCh38
rs371626610 Chr.9: 73168987-73168987 on Build GRCh38
rs3739959 Chr.9: 73169021-73169021 on Build GRCh38
rs754170721 Chr.9: 73169126-73169126 on Build GRCh38

View Full Product Details