Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 13927901-13928170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
UniGene
Hs.567265
Gene Chromosome Location
Chr.16: 13920138-13952348 on Build GRCh38
Transcripts
NM_005236.2 XM_011522424.2 XM_017023043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6271666 c.670G>T p.A224S Chr.16: 13928113-13928113 on Build GRCh38
6906927 c.628G>T p.E210* Chr.16: 13928071-13928071 on Build GRCh38
4681593 c.585-2A>G p.? Chr.16: 13928026-13928026 on Build GRCh38
6957565 c.725G>A p.C242Y Chr.16: 13928168-13928168 on Build GRCh38
1301637 c.628G>A p.E210K Chr.16: 13928071-13928071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764731249 Chr.16: 13928118-13928118 on Build GRCh38
rs969667669 Chr.16: 13928131-13928131 on Build GRCh38
rs753799081 Chr.16: 13928110-13928110 on Build GRCh38
rs1022507611 Chr.16: 13928128-13928128 on Build GRCh38
rs758362908 Chr.16: 13928104-13928105 on Build GRCh38
rs777165799 Chr.16: 13927992-13927992 on Build GRCh38
rs375292949 Chr.16: 13927984-13927984 on Build GRCh38
rs769983967 Chr.16: 13928010-13928010 on Build GRCh38
rs1037934959 Chr.16: 13928069-13928069 on Build GRCh38
rs397509402 Chr.16: 13928132-13928132 on Build GRCh38
rs779737289 Chr.16: 13928129-13928129 on Build GRCh38
rs748246572 Chr.16: 13927993-13927993 on Build GRCh38
rs899417305 Chr.16: 13928142-13928142 on Build GRCh38
rs766656010 Chr.16: 13928064-13928064 on Build GRCh38
rs145138851 Chr.16: 13928095-13928095 on Build GRCh38
rs764003312 Chr.16: 13928098-13928098 on Build GRCh38
rs780428055 Chr.16: 13927978-13927978 on Build GRCh38
rs757128317 Chr.16: 13928116-13928116 on Build GRCh38
rs749895744 Chr.16: 13928126-13928126 on Build GRCh38
rs775194104 Chr.16: 13928074-13928074 on Build GRCh38
rs533954045 Chr.16: 13928013-13928013 on Build GRCh38
rs747350732 Chr.16: 13927979-13927979 on Build GRCh38
rs1011502680 Chr.16: 13928117-13928117 on Build GRCh38
rs943532569 Chr.16: 13927970-13927970 on Build GRCh38
rs568308691 Chr.16: 13927969-13927969 on Build GRCh38
rs889319305 Chr.16: 13927955-13927955 on Build GRCh38
rs763276174 Chr.16: 13928027-13928027 on Build GRCh38
rs766148211 Chr.16: 13928112-13928123 on Build GRCh38

View Full Product Details