Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50581740-50581971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6628414 c.443T>C p.I148T Chr.22: 50581753-50581753 on Build GRCh38
1190501 c.386G>T p.R129L Chr.22: 50581810-50581810 on Build GRCh38
7314054 c.376C>A p.L126I Chr.22: 50581820-50581820 on Build GRCh38
5796569 c.349G>C p.V117L Chr.22: 50581847-50581847 on Build GRCh38
1248242 c.381G>A p.A127A Chr.22: 50581815-50581815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767938046 Chr.22: 50581811-50581811 on Build GRCh38
rs772143163 Chr.22: 50581840-50581840 on Build GRCh38
rs758986822 Chr.22: 50581871-50581871 on Build GRCh38
rs762694630 Chr.22: 50581771-50581771 on Build GRCh38
rs142607228 Chr.22: 50581842-50581842 on Build GRCh38
rs201257544 Chr.22: 50581768-50581768 on Build GRCh38
rs757579942 Chr.22: 50581898-50581898 on Build GRCh38
rs138424307 Chr.22: 50581879-50581879 on Build GRCh38
rs977438817 Chr.22: 50581787-50581787 on Build GRCh38
rs761240597 Chr.22: 50581892-50581892 on Build GRCh38
rs748051557 Chr.22: 50581844-50581844 on Build GRCh38
rs150987240 Chr.22: 50581862-50581862 on Build GRCh38
rs754177972 Chr.22: 50581896-50581896 on Build GRCh38
rs189564179 Chr.22: 50581850-50581850 on Build GRCh38
rs764498952 Chr.22: 50581893-50581893 on Build GRCh38
rs559769048 Chr.22: 50581920-50581920 on Build GRCh38
rs760126717 Chr.22: 50581807-50581807 on Build GRCh38
rs757454951 Chr.22: 50581832-50581832 on Build GRCh38
rs375735904 Chr.22: 50581911-50581911 on Build GRCh38
rs753195116 Chr.22: 50581816-50581816 on Build GRCh38
rs770673756 Chr.22: 50581868-50581868 on Build GRCh38
rs765468048 Chr.22: 50581905-50581905 on Build GRCh38
rs745967029 Chr.22: 50581836-50581836 on Build GRCh38
rs774104710 Chr.22: 50581869-50581869 on Build GRCh38
rs368142650 Chr.22: 50581800-50581800 on Build GRCh38
rs375802036 Chr.22: 50581928-50581928 on Build GRCh38
rs779012297 Chr.22: 50581833-50581833 on Build GRCh38
rs548828466 Chr.22: 50581946-50581946 on Build GRCh38
rs990631675 Chr.22: 50581925-50581925 on Build GRCh38
rs778043765 Chr.22: 50581826-50581826 on Build GRCh38
rs900079936 Chr.22: 50581947-50581947 on Build GRCh38
rs1025908134 Chr.22: 50581834-50581834 on Build GRCh38
rs766046984 Chr.22: 50581773-50581773 on Build GRCh38
rs754126452 Chr.22: 50581831-50581831 on Build GRCh38
rs1054745614 Chr.22: 50581943-50581943 on Build GRCh38
rs750601369 Chr.22: 50581909-50581909 on Build GRCh38
rs371606408 Chr.22: 50581867-50581867 on Build GRCh38
rs761535800 Chr.22: 50581758-50581758 on Build GRCh38
rs769546336 Chr.22: 50581847-50581847 on Build GRCh38
rs756445442 Chr.22: 50581823-50581823 on Build GRCh38
rs753147853 Chr.22: 50581887-50581887 on Build GRCh38

View Full Product Details