Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1361857-1362096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5483984 c.276C>T p.H92H Chr.16: 1361914-1361914 on Build GRCh38
5548321 c.298G>A p.A100T Chr.16: 1361936-1361936 on Build GRCh38
1718978 c.341C>T p.A114V Chr.16: 1362061-1362061 on Build GRCh38
1208435 c.340G>A p.A114T Chr.16: 1362060-1362060 on Build GRCh38
4058421 c.235T>C p.Y79H Chr.16: 1361873-1361873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781472406 Chr.16: 1361971-1361971 on Build GRCh38
rs778178434 Chr.16: 1361889-1361889 on Build GRCh38
rs1032142138 Chr.16: 1362010-1362010 on Build GRCh38
rs200975355 Chr.16: 1362067-1362067 on Build GRCh38
rs201531462 Chr.16: 1361915-1361915 on Build GRCh38
rs764395288 Chr.16: 1361903-1361903 on Build GRCh38
rs752836392 Chr.16: 1362070-1362070 on Build GRCh38
rs201992413 Chr.16: 1361914-1361914 on Build GRCh38
rs376026722 Chr.16: 1362009-1362009 on Build GRCh38
rs768817513 Chr.16: 1361988-1361988 on Build GRCh38
rs774673554 Chr.16: 1361953-1361953 on Build GRCh38
rs149197775 Chr.16: 1361907-1361907 on Build GRCh38
rs748941730 Chr.16: 1361881-1361881 on Build GRCh38
rs901500768 Chr.16: 1361996-1361996 on Build GRCh38
rs765847099 Chr.16: 1361923-1361923 on Build GRCh38
rs769001628 Chr.16: 1361950-1361950 on Build GRCh38
rs1038742616 Chr.16: 1362033-1362033 on Build GRCh38
rs148378749 Chr.16: 1362020-1362020 on Build GRCh38
rs770716391 Chr.16: 1362038-1362038 on Build GRCh38
rs200116947 Chr.16: 1361991-1361991 on Build GRCh38
rs775604993 Chr.16: 1361901-1361901 on Build GRCh38
rs758452962 Chr.16: 1362077-1362077 on Build GRCh38
rs749320662 Chr.16: 1361942-1361942 on Build GRCh38
rs778481117 Chr.16: 1361928-1361928 on Build GRCh38
rs745850911 Chr.16: 1362043-1362043 on Build GRCh38
rs201045559 Chr.16: 1361935-1361935 on Build GRCh38
rs769722097 Chr.16: 1362045-1362045 on Build GRCh38
rs201978812 Chr.16: 1362005-1362005 on Build GRCh38
rs768511134 Chr.16: 1361926-1361926 on Build GRCh38
rs771330629 Chr.16: 1361988-1361992 on Build GRCh38
rs779191841 Chr.16: 1362025-1362025 on Build GRCh38
rs779778381 Chr.16: 1361880-1361880 on Build GRCh38
rs193302849 Chr.16: 1362067-1362069 on Build GRCh38
rs754595108 Chr.16: 1361884-1361884 on Build GRCh38
rs769717288 Chr.16: 1361898-1361898 on Build GRCh38
rs145313679 Chr.16: 1361902-1361902 on Build GRCh38
rs371160615 Chr.16: 1362015-1362015 on Build GRCh38
rs374558065 Chr.16: 1361967-1361967 on Build GRCh38
rs371195304 Chr.16: 1361958-1361958 on Build GRCh38
rs752300611 Chr.16: 1361969-1361969 on Build GRCh38
rs770325965 Chr.16: 1361936-1361936 on Build GRCh38
rs370815540 Chr.16: 1361959-1361959 on Build GRCh38
rs753385216 Chr.16: 1361927-1361927 on Build GRCh38
rs926216364 Chr.16: 1361962-1361962 on Build GRCh38
rs367661016 Chr.16: 1362060-1362060 on Build GRCh38
rs139385234 Chr.16: 1361940-1361940 on Build GRCh38
rs756455159 Chr.16: 1362016-1362016 on Build GRCh38
rs137852884 Chr.16: 1362053-1362053 on Build GRCh38
rs76594024 Chr.16: 1361893-1361893 on Build GRCh38
rs757581909 Chr.16: 1361932-1361932 on Build GRCh38
rs137852885 Chr.16: 1361954-1361954 on Build GRCh38
rs756586617 Chr.16: 1361979-1361979 on Build GRCh38
rs543804571 Chr.16: 1361908-1361908 on Build GRCh38
rs758027840 Chr.16: 1361970-1361970 on Build GRCh38
rs756056220 Chr.16: 1361922-1361922 on Build GRCh38
rs750277954 Chr.16: 1361917-1361917 on Build GRCh38
rs763127645 Chr.16: 1361926-1361926 on Build GRCh38
rs745994739 Chr.16: 1361897-1361897 on Build GRCh38
rs2072987 Chr.16: 1362004-1362004 on Build GRCh38
rs960073699 Chr.16: 1362013-1362013 on Build GRCh38
rs772946278 Chr.16: 1362001-1362001 on Build GRCh38
rs776376435 Chr.16: 1362041-1362041 on Build GRCh38
rs141533972 Chr.16: 1362047-1362047 on Build GRCh38
rs112358354 Chr.16: 1361976-1361976 on Build GRCh38
rs375764145 Chr.16: 1362026-1362026 on Build GRCh38
rs184790515 Chr.16: 1361992-1361992 on Build GRCh38
rs762664496 Chr.16: 1362011-1362011 on Build GRCh38
rs143383133 Chr.16: 1361933-1361933 on Build GRCh38
rs367754651 Chr.16: 1361983-1361983 on Build GRCh38
rs143316080 Chr.16: 1362019-1362019 on Build GRCh38
rs764763952 Chr.16: 1362008-1362008 on Build GRCh38
rs113058618 Chr.16: 1362014-1362014 on Build GRCh38
rs772324860 Chr.16: 1361995-1361995 on Build GRCh38
rs774017237 Chr.16: 1362051-1362051 on Build GRCh38
rs138297609 Chr.16: 1362064-1362064 on Build GRCh38
rs193302847 Chr.16: 1362037-1362037 on Build GRCh38
rs747362604 Chr.16: 1361892-1361892 on Build GRCh38
rs900341724 Chr.16: 1361997-1361997 on Build GRCh38
rs763678034 Chr.16: 1362048-1362048 on Build GRCh38
rs777991598 Chr.16: 1362028-1362028 on Build GRCh38
rs147048228 Chr.16: 1362059-1362059 on Build GRCh38
rs767072821 Chr.16: 1362006-1362006 on Build GRCh38
rs371376391 Chr.16: 1362007-1362007 on Build GRCh38
rs559549488 Chr.16: 1361968-1361968 on Build GRCh38

View Full Product Details